Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorotoluene, 98%

Catalog No. p-7044115
Change view
Click to view available options
Quantity:
100 g
500 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0467222 100 g
AAL0467236 500 g
AAL046720B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0467222 Supplier Thermo Scientific Chemicals Supplier No. L0467222
Add to Cart

CAS: 106-43-4 | C7H7Cl | 126.58 g/mol

4-Chlorotoluene is used for the preparation model gas of 4-chlorotoulene by passing synthesized air into liquid 4-chlorotoulene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorotoluene is used for the preparation model gas of 4-chlorotoulene by passing synthesized air into liquid 4-chlorotoulene.

Solubility
Soluble in water (0.04 g/L at 20oc).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-43-4
Molecular Formula C7H7Cl
Molecular Weight (g/mol) 126.58
MDL Number MFCD00000631
InChI Key NPDACUSDTOMAMK-UHFFFAOYSA-N
Synonym 4-chlorotoluene, p-chlorotoluene, benzene, 1-chloro-4-methyl, p-tolyl chloride, toluene, p-chloro, 1-methyl-4-chlorobenzene, 4-chloro-1-methylbenzene, monochlorotoluene, para-chlorotoluene, unii-q8r236h42n
PubChem CID 7810
ChEBI CHEBI:34401
IUPAC Name 1-chloro-4-methylbenzene
SMILES CC1=CC=C(Cl)C=C1

Specifications

Melting Point 7°C to 9°C
Density 1.069
Quantity 100 g
Odor Aromatic
Refractive Index 1.52
UN Number UN2238
Beilstein 1903635
Merck Index 14,2171
Boiling Point 161°C to 163°C
Solubility Information Soluble in water (0.04g/L at 20oc).
Formula Weight 126.59
Percent Purity 98%
Chemical Name or Material 4-Chlorotoluene
Show More Show Less
Hazard Category H226-H317-H332
Hazard Statement GHS H Statement
H226-H332
Flammable liquid and vapor.
Harmful if inhaled.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P271-P272-P280-P303+P361+P353-P304+P340-P312-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CHLOROTOLUENES
EINECSNumber 203-397-0
RTECSNumber XS9010000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.