Learn More
Sigma Organic Chemistry 4-Cyano-1H-pyrrole-2-ca 2 | 25mg | MFCD09991904
Supplier: Sigma Organic Chemistry CDS00689625MG

4-Cyano-1H-pyrrole-2-ca 2, 25mg
About this Item:
MDL #: MFCD09991904
Molecular Weight: 136.11
UNSPSC Code: 12352200
Chemical Formula: C6H4N2O2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.