Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Cyano-4-[(dodecylsulfanylthiocarbonyl)sulfanyl]pentanol

Catalog No. 7601101G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-011-01G 1 g
76-011-05G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 76-011-01G Supplier Sigma Aldrich Supplier No. 7601101G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reversible addition-fragmentation chain transfer (RAFT) polymerisation is used to produce narrow polydispersity polymers of complex architecture. 4-Cyano-4-[(dodecylsulfanylthiocarbonyl)sulfanyl]pentanol is an hydroxyl functionalised trithiocarbonate RAFT agent.It is used as a dual initiator for RAFT and ring opening polymerisation to form well-defined block copolymers. They have good control of polymerisation for a wide range of vinyl monomers.Initiator for RAFT and ring opening polymerisation of block copolymers derived from vinyl monomers of styrene, metacryalte, acrylamide and cyclic monomers.

Specifications

Molecular Formula C19H35NOS3
Linear Formula C19H35NOS3
MDL Number MFCD22417215
Quantity 1 g
Molecular Weight (g/mol) 389.68
Percent Purity ≥96.5% (HPLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.