Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyano-4'-hydroxybiphenyl 98.0+%, TCI America™
SDP

Catalog No. H052325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H052325G 25 g
H05235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H052325G Supplier TCI America Supplier No. H052325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19812-93-2
Molecular Formula C13H9NO
Molecular Weight (g/mol) 195.221
MDL Number MFCD00059625
InChI Key ZRMIETZFPZGBEB-UHFFFAOYSA-N
Synonym 4-cyano-4'-hydroxybiphenyl, 4'-hydroxy-4-biphenylcarbonitrile, 4-4-hydroxyphenyl benzonitrile, 4'-hydroxybiphenyl-4-carbonitrile, 4'-cyano-4-hydroxybiphenyl, 4'-hydroxy-1,1'-biphenyl-4-carbonitrile, 4'-cyano-4-biphenylol, 4-cyano-4'-hydroxy-biphenyl, 4'-hydroxy-4-cyanobiphenyl, 4-hydroxy-4-biphenylcarbonitrile
PubChem CID 140610
IUPAC Name 4-(4-hydroxyphenyl)benzonitrile
SMILES C1=CC(=CC=C1C#N)C2=CC=C(C=C2)O

Specifications

Melting Point 198°C
Color White-Yellow
Quantity 25 g
Formula Weight 195.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyano-4′-hydroxybiphenyl
EINECSNumber (4)-1822
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.