Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyano-4'-methylbiphenyl 98.0+%, TCI America™
SDP

Catalog No. C174125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C174125G 25 g
C17415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C174125G Supplier TCI America Supplier No. C174125G

Chemical Identifiers

CAS 50670-50-3
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.249
MDL Number MFCD03703444
InChI Key QIBWMVSMTSYUSK-UHFFFAOYSA-N
PubChem CID 170889
IUPAC Name 4-(4-methylphenyl)benzonitrile
SMILES CC1=CC=C(C=C1)C2=CC=C(C=C2)C#N

Specifications

Melting Point 110°C
Color White
Quantity 25 g
Synonym 4′-Methyl-4-biphenylcarbonitrile
Formula Weight 193.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyano-4′-methylbiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.