Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyano-4-phenylcyclohexanone, 97%

Catalog No. aal0528106
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0528106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0528106 Supplier Thermo Scientific Chemicals Supplier No. L0528106
Only null left
Add to Cart

CAS: 25115-74-6 | C13H13NO | 199.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25115-74-6
Molecular Formula C13H13NO
Molecular Weight (g/mol) 199.25
MDL Number MFCD00044816
InChI Key GKXOABVSZWCJJK-UHFFFAOYSA-N
Synonym 4-cyano-4-phenylcyclohexanone, 4-oxo-1-phenylcyclohexanecarbonitrile, 4-oxo-1-phenylcyclohexanenitrile, cyclohexanecarbonitrile, 4-oxo-1-phenyl, 4-oxo-1-phenyl-cyclohexanecarbonitrile, 4-oxo-1-phenyl-1-cyclohexanecarbonitrile, acmc-1ciss, 4-cyano-4phenylcyclohexanone, 4-phenyl 4-cyanocylohexanone, 4-phenyl 4-cyanocyclohexanone
PubChem CID 91282
IUPAC Name 4-oxo-1-phenylcyclohexane-1-carbonitrile
SMILES O=C1CCC(CC1)(C#N)C1=CC=CC=C1

Specifications

Melting Point 112°C to 118°C
Quantity 5 g
Beilstein 2645593
Formula Weight 199.24
Percent Purity 98%
Chemical Name or Material 4-Cyano-4-phenylcyclohexanone
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 246-631-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.