Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyanobenzyl Bromide 98.0+%, TCI America™
SDP

Catalog No. C1398250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1398250G 250 g
C139825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1398250G Supplier TCI America Supplier No. C1398250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17201-43-3
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.047
MDL Number MFCD00001829
InChI Key UMLFTCYAQPPZER-UHFFFAOYSA-N
Synonym 4-cyanobenzyl bromide, 4-bromomethyl benzonitrile, p-cyanobenzyl bromide, alpha-bromo-p-tolunitrile, benzonitrile, 4-bromomethyl, 4-bromomethyl-benzonitrile, alpha-bromo-p-toluonitrile, p-bromomethyl benzonitrile, 4-cyano benzyl bromide, benzonitrile, p-bromomethyl
PubChem CID 86996
IUPAC Name 4-(bromomethyl)benzonitrile
SMILES C1=CC(=CC=C1CBr)C#N

Specifications

Melting Point 116°C
Color White
Boiling Point 143°C
Quantity 250 g
UN Number 3261
Formula Weight 196.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyanobenzyl Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.