Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyanobiphenyl 98.0+%, TCI America™
SDP

Catalog No. C129925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C129925G 25 g
C12995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C129925G Supplier TCI America Supplier No. C129925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2920-38-9
Molecular Formula C13H9N
Molecular Weight (g/mol) 179.22
MDL Number MFCD00001821
InChI Key BPMBNLJJRKCCRT-UHFFFAOYSA-N
Synonym 4-cyanobiphenyl, 1,1'-biphenyl-4-carbonitrile, 4-biphenylcarbonitrile, p-phenylbenzonitrile, p-cyanobiphenyl, biphenyl-4-carbonitrile, 4-phenylbenzenecarbonitrile, 4-biphenylcarboxylic acid nitrile, 4-cyanodiphenyl
PubChem CID 18021
IUPAC Name [1,1'-biphenyl]-4-carbonitrile
SMILES N#CC1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 87°C
Color White-Yellow
Boiling Point 193°C
Quantity 25 g
UN Number 3439
Formula Weight 179.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyanobiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.