Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyanopyridine N-Oxide 98.0+%, TCI America™
SDP

Catalog No. C076525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C076525G 25 g
C0765500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C076525G Supplier TCI America Supplier No. C076525G
 Restricted Item
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14906-59-3
Molecular Formula C6H4N2O
Molecular Weight (g/mol) 120.11
MDL Number MFCD00006205
InChI Key QNCSFBSIWVBTHE-UHFFFAOYSA-N
Synonym Isonicotinonitrile N-Oxide
PubChem CID 84684
IUPAC Name 4-cyanopyridin-1-ium-1-olate
SMILES [O-][N+]1=CC=C(C=C1)C#N

Specifications

Melting Point 230°C
Color Yellow
Quantity 25 g
Formula Weight 120.11
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyanopyridine N-Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.