Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyclohexyl N-(tert-Butoxycarbonyl)-L-aspartate 98.0+%, TCI America™
SDP

Catalog No. C253525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C253525G 25 g
C25355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C253525G Supplier TCI America Supplier No. C253525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 73821-95-1
Molecular Formula C15H25NO6
Molecular Weight (g/mol) 315.366
MDL Number MFCD00061996
InChI Key NLPQIWFEEKQBBN-NSHDSACASA-N
Synonym boc-asp ochx-oh, boc-asp ochex-oh, boc-l-aspartic acid 4-cyslohexyl ester, n-boc-l-aspartic acid 4-cyclohexyl ester, 2s-2-tert-butoxycarbonyl amino-4-cyclohexyloxy-4-oxobutanoic acid, boc-asp o-chex, ambotzbaa1074, pubchem12097, 4-cyclohexyl n-boc-l-aspartate, boc-l-aspartic acid b-cyclohexyl ester
PubChem CID 7009416
IUPAC Name (2S)-4-cyclohexyloxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)OC1CCCCC1)C(=O)O

Specifications

Melting Point 96°C
Color White
Quantity 25 g
Formula Weight 315.37
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyclohexyl N-(tert-Butoxycarbonyl)-L-aspartate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.