Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Decanone 97.0+%, TCI America™
SDP

Catalog No. D153510ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D153510ML 10 mL
D153525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D153510ML Supplier TCI America Supplier No. D153510ML

Chemical Identifiers

CAS 624-16-8
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD00009542
InChI Key MKJDUHZPLQYUCB-UHFFFAOYSA-N
Synonym 4-decanone, hexyl propyl ketone, propyl hexyl ketone, unii-502v1w8kd4, n-propyl n-hexyl ketone, acmc-209n3f, mkjduhzplqyucb-uhfffaoysa, unii-3z25263nt0 component
PubChem CID 12203
IUPAC Name decan-4-one
SMILES CCCCCCC(=O)CCC

Specifications

Color Yellow
Boiling Point 90°C
Quantity 10 mL
Formula Weight 156.27
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Decanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.