Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Diethylaminobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. D249325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D249325G 25 g
D24935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D249325G Supplier TCI America Supplier No. D249325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5429-28-7
Molecular Formula C11H15NO2
Molecular Weight (g/mol) 193.246
MDL Number MFCD00002538
InChI Key LNYTUARMNSFFBE-UHFFFAOYSA-N
Synonym 4-diethylamino benzoic acid, benzoic acid, 4-diethylamino, 4-n,n-diethylaminobenzoic acid, p-diethylamino benzoic acid, 4-diethylaminobenzoicacid, acmc-209lfa, 4-diethylamino benzoic, p-diethylaminobenzoic acid, 4-diethylamino benzoicacid
PubChem CID 79480
IUPAC Name 4-(diethylamino)benzoic acid
SMILES CCN(CC)C1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 196°C
Color White-Yellow
Quantity 25 g
Formula Weight 193.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Diethylaminobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.