Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Difluoromethoxy)benzenesulfonamide 98.0+%, TCI America™
SDP

Catalog No. D47781G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47781G 1 g
D47785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47781G Supplier TCI America Supplier No. D47781G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 874781-09-6
Molecular Formula C7H7F2NO3S
Molecular Weight (g/mol) 223.19
MDL Number MFCD03407973
InChI Key BBBQHNZWCPUJNW-UHFFFAOYSA-N
PubChem CID 24820749
IUPAC Name 4-(difluoromethoxy)benzene-1-sulfonamide
SMILES NS(=O)(=O)C1=CC=C(OC(F)F)C=C1

Specifications

Melting Point 113°C
Color White-Yellow
Quantity 1 g
Formula Weight 223.19
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Difluoromethoxy)benzenesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.