Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Difluoromethoxy)benzoic Acid 98.0+%, TCI America™
SDP

Catalog No. D47621G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47621G 1 g
D47625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47621G Supplier TCI America Supplier No. D47621G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4837-20-1
Molecular Formula C8H5F2O3
Molecular Weight (g/mol) 187.12
MDL Number MFCD00221454
InChI Key BSNNYLYELGBSBA-UHFFFAOYSA-M
Synonym 4-difluoromethoxy benzoic acid, 4-difluoromethoxy-benzoic acid, benzoic acid, 4-difluoromethoxy, pubchem3933, acmc-20a19a, p-difluoromethoxy benzoic acid, alpha,alpha-difluoro-p-anisic acid, 4-difluoromethyl oxy benzoic acid
PubChem CID 737360
IUPAC Name 4-(difluoromethoxy)benzoate
SMILES [O-]C(=O)C1=CC=C(OC(F)F)C=C1

Specifications

Melting Point 170°C
Color White-Yellow
Quantity 1 g
Formula Weight 188.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Difluoromethoxy)benzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.