Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Dimethylaminoacetophenone 98.0+%, TCI America™
SDP

Catalog No. D157525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D157525G 25 g
D15755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D157525G Supplier TCI America Supplier No. D157525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2124-31-4
Molecular Formula C10H13NO
Molecular Weight (g/mol) 163.22
MDL Number MFCD00017246
InChI Key HUDYANRNMZDQGA-UHFFFAOYSA-N
PubChem CID 75037
IUPAC Name 1-[4-(dimethylamino)phenyl]ethan-1-one
SMILES CN(C)C1=CC=C(C=C1)C(C)=O

Specifications

Melting Point 105°C
Color White-Yellow
Quantity 25 g
Formula Weight 163.22
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Dimethylaminoacetophenone
EINECSNumber (3)-2939
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.