Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Dimethylaminoazobenzene-4'-carboxylic Acid 97.0+%, TCI America™
SDP

Catalog No. M042325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M042325G 25 g
M04235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M042325G Supplier TCI America Supplier No. M042325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6268-49-1
Molecular Formula C15H15N3O2
Molecular Weight (g/mol) 269.30
MDL Number MFCD00058988
InChI Key WCKQPPQRFNHPRJ-UHFFFAOYSA-N
Synonym p-Methyl Red, Para Methyl Red
PubChem CID 22650
IUPAC Name 4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoic acid
SMILES CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)C(O)=O

Specifications

Color Red-Yellow
Quantity 25 g
Formula Weight 269.30
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Dimethylaminoazobenzene-4′-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.