Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Dimethylaminobenzoic acid, 98+%, Thermo Scientific™

Catalog No. AAA1433830
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1433830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1433830 Supplier Thermo Scientific Chemicals Supplier No. A1433830

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Dimethylaminobenzoic acid a substituted benzoic acid that acts as an inhibitor of ultraviolet mediated damage to skin. It is a biotransformation product of 2-ethylhexyl 4- (N,N-dimethylaminobenzo)benzoate(EDP), a widely used UV filters in sunscreen cosmetics and other cosmetic products.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-84-1
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00002537
InChI Key YDIYEOMDOWUDTJ-UHFFFAOYSA-N
Synonym 4-dimethylamino benzoic acid, benzoic acid, 4-dimethylamino, benzoic acid, p-dimethylamino, p-dimethylaminobenzoic acid, n,n-dimethyl-p-aminobenzoic acid, n,n-dimethyl-4-aminobenzoic acid, p-n,n-dimethylamino benzoic acid, p-dimethylamino benzoic acid, unii-d1ma908ev0, 4-n,n-dimethylamino benzoic acid
PubChem CID 12092
IUPAC Name 4-(dimethylamino)benzoic acid
SMILES CN(C)C1=CC=C(C=C1)C(=O)O

Specifications

Melting Point ∼240°C (decomposition)
Flash Point 170°C (338°F)
Odor Odorless
Quantity 250 g
Beilstein 386867
Merck Index 14,3232
Solubility Information Insoluble in water.
Formula Weight 165.19
Percent Purity ≥98%
Chemical Name or Material 4-Dimethylaminobenzoic acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-615-8
RTECSNumber DG8735000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.