CAS RN 619-84-1
4-Dimethylaminobenzoic acid, 98%, Thermo Scientific Chemicals
CAS: 619-84-1 Molecular Formula: C9H11NO2 Molecular Weight (g/mol): 165.19 MDL Number: MFCD00002537 InChI Key: YDIYEOMDOWUDTJ-UHFFFAOYSA-N Synonym: 4-dimethylamino benzoic acid,benzoic acid, 4-dimethylamino,benzoic acid, p-dimethylamino,p-dimethylaminobenzoic acid,n,n-dimethyl-p-aminobenzoic acid,n,n-dimethyl-4-aminobenzoic acid,p-n,n-dimethylamino benzoic acid,p-dimethylamino benzoic acid,unii-d1ma908ev0,4-n,n-dimethylamino benzoic acid PubChem CID: 12092 IUPAC Name: 4-(dimethylamino)benzoic acid SMILES: CN(C)C1=CC=C(C=C1)C(=O)O
4-Dimethylaminobenzoic Acid 98.0+%, TCI America™
CAS: 619-84-1 Molecular Formula: C9H11NO2 Molecular Weight (g/mol): 165.192 MDL Number: MFCD00002537 InChI Key: YDIYEOMDOWUDTJ-UHFFFAOYSA-N Synonym: 4-dimethylamino benzoic acid,benzoic acid, 4-dimethylamino,benzoic acid, p-dimethylamino,p-dimethylaminobenzoic acid,n,n-dimethyl-p-aminobenzoic acid,n,n-dimethyl-4-aminobenzoic acid,p-n,n-dimethylamino benzoic acid,p-dimethylamino benzoic acid,unii-d1ma908ev0,4-n,n-dimethylamino benzoic acid PubChem CID: 12092 IUPAC Name: 4-(dimethylamino)benzoic acid SMILES: CN(C)C1=CC=C(C=C1)C(=O)O
Sigma Aldrich 1,2:5,6-Di-O-Isopropylidene-Alpha-D-Glucofuranose
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
4-Dimethylaminobenzoic acid, 98+%, Thermo Scientific™
CAS: 619-84-1 Molecular Formula: C9H11NO2 Molecular Weight (g/mol): 165.192 MDL Number: MFCD00002537 InChI Key: YDIYEOMDOWUDTJ-UHFFFAOYSA-N Synonym: 4-dimethylamino benzoic acid,benzoic acid, 4-dimethylamino,benzoic acid, p-dimethylamino,p-dimethylaminobenzoic acid,n,n-dimethyl-p-aminobenzoic acid,n,n-dimethyl-4-aminobenzoic acid,p-n,n-dimethylamino benzoic acid,p-dimethylamino benzoic acid,unii-d1ma908ev0,4-n,n-dimethylamino benzoic acid PubChem CID: 12092 IUPAC Name: 4-(dimethylamino)benzoic acid SMILES: CN(C)C1=CC=C(C=C1)C(=O)O
MP Biomedicals, Inc p-Dimethylaminobenzoic Acid, MP Biomedicals
CAS: 619-84-1 Molecular Formula: C9H11NO2 Molecular Weight (g/mol): 165.192 InChI Key: YDIYEOMDOWUDTJ-UHFFFAOYSA-N Synonym: 4-dimethylamino benzoic acid,benzoic acid, 4-dimethylamino,benzoic acid, p-dimethylamino,p-dimethylaminobenzoic acid,n,n-dimethyl-p-aminobenzoic acid,n,n-dimethyl-4-aminobenzoic acid,p-n,n-dimethylamino benzoic acid,p-dimethylamino benzoic acid,unii-d1ma908ev0,4-n,n-dimethylamino benzoic acid PubChem CID: 12092 IUPAC Name: 4-(dimethylamino)benzoic acid SMILES: CN(C)C1=CC=C(C=C1)C(=O)O