Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Diphenylamino)benzeneboronic acid, 98%

Catalog No. AAH5290203
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5290203 1 g
AAH52902MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5290203 Supplier Thermo Scientific Chemicals Supplier No. H5290203

CAS: 201802-67-7 | C18H16BNO2 | 289.141 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 201802-67-7
Molecular Formula C18H16BNO2
Molecular Weight (g/mol) 289.141
MDL Number MFCD06798117
InChI Key TWWQCBRELPOMER-UHFFFAOYSA-N
Synonym 4-diphenylamino phenylboronic acid, triphenylamine-4-boronic acid, 4-diphenylamino phenyl boronic acid, 4-diphenylamino benzeneboronic acid, 4-n,n-diphenylamino phenylboronic acid, boronic acid, b-4-diphenylamino phenyl, 4-n-diphenylamino phenylboronic acid, 4-diphenylamino phenylboronicacid, boronic acid, 4-diphenylamino phenyl
PubChem CID 12166934
IUPAC Name [4-(N-phenylanilino)phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=CC=C3)(O)O

Specifications

Melting Point 110°C to 115°C
Quantity 1 g
Formula Weight 289.14
Percent Purity 98%
Chemical Name or Material 4-(Diphenylamino)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.