Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Dodecyloxyphthalonitrile 99.0+%, TCI America™
SDP

Catalog No. D22371G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22371G 1 g
D22375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D22371G Supplier TCI America Supplier No. D22371G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 161082-75-3
Molecular Formula C20H28N2O
Molecular Weight (g/mol) 312.457
MDL Number MFCD00191413
InChI Key SWVGQFFVXRAJRQ-UHFFFAOYSA-N
Synonym 1,2-Dicyano-4-dodecyloxybenzene
PubChem CID 44630254
IUPAC Name 4-dodecoxybenzene-1,2-dicarbonitrile
SMILES CCCCCCCCCCCCOC1=CC(=C(C=C1)C#N)C#N

Specifications

Melting Point 62°C
Quantity 1 g
Formula Weight 312.46
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Dodecyloxyphthalonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.