Learn More
Cayman Chemical 4-EAPBhydrochlorIde 1mg
SDP
Supplier:  Cayman Chemical 90021021 MG
Encompass_Preferred
An oxindole alkaloid with diverse biological activities induces activation of PXR in a reporter assay using DPX2 cells expressing the human receptor (EC50 0.7656 UM) a positive modulator of M1 mAChRs and 5-HT2 receptor (EC50s 9.92 and 14.5 UM for the rat receptors respectively) active against S. aureus and B. subtilis (MICs 408 and 679 UM respectively) inhibits the proliferation of CCRF CEM C7H2 lymphoblastic leukemia cells at 100 UM
Catalog No. 50-289-2943
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.