Learn More
Medchemexpress LLC 4-epioxytetracycline | 14206-58-7 | MFCD00021181 | >98.0% | 460.43 | C22H24N2O9 | 5MG
Supplier: Medchemexpress LLC HY1259475MG
4-Epioxytetracycline is an analytical reference standard of 4-epi-oxytetracycline, a degradation product of the antibiotic oxytetracycline (OTC). It is supplied for research and analytical applications, including chromatographic and mass spectrometric method development, validation, and identification of residues and metabolites in environmental and biological samples.
- Analytical standard for method development and validation.
- Intended for residue and metabolite identification in environmental and biological samples.
- High purity (≥98%) suitable for chromatographic and mass spectrometric analysis.
- Available in small, laboratory-scale quantities for analytical use.
- Refer to the safety data sheet for storage and handling recommendations.