Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethoxy-2-methylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. E11731G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11731G 1 g
E1173-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11731G Supplier TCI America Supplier No. E11731G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 313545-31-2
Molecular Formula C9H13BO3
Molecular Weight (g/mol) 180.01
MDL Number MFCD04115659
InChI Key MBEQXBKSJCOKJF-UHFFFAOYSA-N
PubChem CID 16217476
IUPAC Name (4-ethoxy-2-methylphenyl)boronic acid

Specifications

Melting Point 168°C
Color White
Quantity 1 g
Synonym 4-ethoxy-2-methylbenzeneboronic acid, 4-ethoxy-2-methylphenylboronicacid, 4-ethoxy-2-methylphenyl boronic acid, 5-ethoxytoluene-2-boronic acid, 4-ethoxy-2-methyl-phenyl boronic acid, boronic acid,b-4-ethoxy-2-methylphenyl, acmc-1aexl, 2-methyl-4-ethoxybenzene boronic acid, contains varying amounts of anhydride
SMILES B(C1=C(C=C(C=C1)OCC)C)(O)O
Formula Weight 180.01
Physical Form Crystalline Powder
Chemical Name or Material 4-Ethoxy-2-methylphenylboronic Acid (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.