Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Ethoxycarbonyl)benzeneboronic acid pinacol ester, 97%, Thermo Scientific™

Catalog No. AAH5583606
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5583606 5 g
AAH55836MD 250 mg
AAH5583603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5583606 Supplier Thermo Scientific Chemicals Supplier No. H5583606

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 195062-62-5
Molecular Formula C15H21BO4
Molecular Weight (g/mol) 276.139
MDL Number MFCD03453659
InChI Key NCVIYKCFTYSAGN-UHFFFAOYSA-N
Synonym ethyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 4-ethoxycarbonylphenylboronic acid pinacol ester, 4-ethoxycarbonylphenylboronic acid, pinacol ester, 4-carboethoxyphenylboronic acid pinacol ester, ethyl 4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, ethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 4-ethoxycarbonyl benzeneboronic acid pinacol ester, 2-4-ethoxycarbonyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid ethyl ester
PubChem CID 2734618
IUPAC Name ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C(=O)OCC

Specifications

Color Colorless
Density 1.052g/mL
Boiling Point 330°C
Refractive Index 1.5000
Quantity 5 g
Formula Weight 276.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-(Ethoxycarbonyl)benzeneboronic acid pinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.