Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-ethylnitrobenzene, 99+%, Thermo Scientific™

Catalog No. AC118751000
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC118751000 100 mL
AC118755000 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC118751000 Supplier Thermo Scientific Chemicals Supplier No. 118751000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-12-9
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00007385
InChI Key RESTWAHJFMZUIZ-UHFFFAOYSA-N
Synonym 4-ethylnitrobenzene, benzene, 1-ethyl-4-nitro, p-ethylnitrobenzene, p-nitroethylbenzene, p-nitrophenylethane, benzene,1-ethyl-4-nitro, 4-ethyl-1-nitrobenzene, 1-ethyl-4-nitro-benzene, ccris 3100, 4-nitroethylbenzene
PubChem CID 7480
IUPAC Name 1-ethyl-4-nitrobenzene
SMILES CCC1=CC=C(C=C1)[N+]([O-])=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color Yellow
Density 1.118g/mL
Infrared Spectrum Authentic
Assay Percent Range 99+%
Refractive Index 1.5445 to 1.5465 (20°C, 589nm)
Linear Formula C2H5C6H4NO2
Quantity 100 mL
Beilstein 05, 358
Formula Weight 151.16
Percent Purity ≥99.0%
Physical Form Liquid
Chemical Name or Material 4-Ethylnitrobenzene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Toxic if inhaled.
Toxic if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.