Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethyloctanoic acid, 97%

Catalog No. AAB2222009
Change view
Click to view available options
Quantity:
2.5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2222009 10 g
AAB2222088 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2222009 Supplier Thermo Scientific Chemicals Supplier No. B2222009
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 16493-80-4 | C10H20O2 | 172.268 g/mol

Five sulfur-containing flavor compounds were synthesized by the reaction of 4-ethyloctanoyl chloride with sulfur-containing alcohols or mercaptans. 4-ethyloctanoic acid is identified from flue-cured Virginia tobacco. Mainly used as daily flavor,food flavor,industrial flavor, nutrient, stabilizers, surfactants and emulsifiers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Five sulfur-containing flavor compounds were synthesized by the reaction of 4-ethyloctanoyl chloride with sulfur-containing alcohols or mercaptans. 4-ethyloctanoic acid is identified from flue-cured Virginia tobacco. Mainly used as daily flavor,food flavor,industrial flavor, nutrient, stabilizers, surfactants and emulsifiers.

Solubility
Slightly soluble in water(0.13 mg/mL). Soluble in hexane

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16493-80-4
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.268
MDL Number MFCD00506494
InChI Key PWKJMPFEQOHBAC-UHFFFAOYSA-N
Synonym octanoic acid, 4-ethyl, 4-ethylcaprylic acid, 4-ethyl octanoic acid, acmc-1bvq5, 4-ethyloctanoicacid, ksc175o1l, 4-ethyloctanoic acid, fg
PubChem CID 61840
IUPAC Name 4-ethyloctanoic acid
SMILES CCCCC(CC)CCC(=O)O

Specifications

Density 0.908
Boiling Point 142°C to 143°C (10 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.439
Quantity 10 g
Solubility Information Slightly soluble in water(0.13mg/mL). Soluble in hexane
Formula Weight 172.27
Percent Purity 97%
Chemical Name or Material 4-Ethyloctanoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 240-560-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.