Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethylphenylacetonitrile, 97%

Catalog No. AAL1085503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1085503 1 g
AAL1085506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1085503 Supplier Thermo Scientific Chemicals Supplier No. L1085503
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 51632-28-1 | C10H11N | 145.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51632-28-1
Molecular Formula C10H11N
Molecular Weight (g/mol) 145.21
MDL Number MFCD00040893
InChI Key NCPKDFDQDZMXCO-UHFFFAOYSA-N
Synonym 4-ethylphenylacetonitrile, 2-4-ethylphenyl acetonitrile, 4-ethylphenyl acetonitrile, benzeneacetonitrile, 4-ethyl, 2-4-ethylphenyl ethanenitrile, 4-ethylbenzylcyanide, p-ethylphenylacetonitrile, acmc-1ao1h
PubChem CID 142867
IUPAC Name 2-(4-ethylphenyl)acetonitrile
SMILES CCC1=CC=C(CC#N)C=C1

Specifications

Density 0.97
Boiling Point 110°C (1 mmHg)
Refractive Index 1.5164
Quantity 1 g
UN Number UN3276
Beilstein 2613187
Formula Weight 145.21
Percent Purity 97%
Chemical Name or Material 4-Ethylphenylacetonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.