Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethylpyridine 98.0+%, TCI America™
SDP

Catalog No. E0170500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0170500ML 500 mL
E017025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0170500ML Supplier TCI America Supplier No. E0170500ML

Chemical Identifiers

CAS 536-75-4
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
MDL Number MFCD00006450
InChI Key VJXRKZJMGVSXPX-UHFFFAOYSA-N
Synonym pyridine, 4-ethyl, .gamma.-ethylpyridine, 4-ethyl pyridine, unii-3kw0se2tl5, 4-ethyl-pyridine, pyridine, c1-3-alkyl derivs., 3kw0se2tl5, gamma-ethylpyridine, c1-c3 alkylpyridine, 4ethylpyridine
PubChem CID 10822
IUPAC Name 4-ethylpyridine
SMILES CCC1=CC=NC=C1

Specifications

Color Yellow
Boiling Point 165°C
Quantity 500 mL
UN Number 1993
Formula Weight 107.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Ethylpyridine
EINECSNumber (5)-3683&(9)-0222
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.