Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethynylpyridine 98.0+%, TCI America™
SDP

Catalog No. E0561100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0561100MG 100 mg
E0561500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0561100MG Supplier TCI America Supplier No. E0561100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2510-22-7
Molecular Formula C7H5N
Molecular Weight (g/mol) 103.124
MDL Number MFCD04038420
InChI Key FDEDJRHULYIJOR-UHFFFAOYSA-N
Synonym 4-Pyridylacetylene
PubChem CID 642801
IUPAC Name 4-ethynylpyridine
SMILES C#CC1=CC=NC=C1

Specifications

Melting Point 98°C
Color White-Yellow
Quantity 100 mg
Formula Weight 103.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Ethynylpyridine
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.