Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Fluoro-3'-(trifluoromethyl)acetophenone 97.0+%, TCI America™
SDP

Catalog No. F08081G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08081G 1 g
F08085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08081G Supplier TCI America Supplier No. F08081G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 208173-24-4
Molecular Formula C9H6F4O
Molecular Weight (g/mol) 206.14
MDL Number MFCD00061252
InChI Key SYFHRXQPXHETEF-UHFFFAOYSA-N
Synonym 4'-fluoro-3'-trifluoromethyl acetophenone, 1-4-fluoro-3-trifluoromethyl phenyl ethanone, 4-fluoro-3-trifluoromethyl acetophenone, 4'-fluoro-3'-trifluoromethylacetophenone, 1-4-fluoro-3-trifluoromethyl phenyl ethan-1-one, 5-acetyl-2-fluorobenzotrifluoride, ethanone, 1-4-fluoro-3-trifluoromethyl phenyl, 1-acetyl-4-fluoro-3-trifluoromethyl benzene, pubchem4321
PubChem CID 605678
IUPAC Name 1-[4-fluoro-3-(trifluoromethyl)phenyl]ethan-1-one
SMILES CC(=O)C1=CC=C(F)C(=C1)C(F)(F)F

Specifications

Color Colorless
Quantity 1 g
Formula Weight 206.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4′-Fluoro-3′-(trifluoromethyl)acetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.