Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-3-(trifluoromethyl)benzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog No. AAH6207103
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6207103 1 g
AAH62071MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6207103 Supplier Thermo Scientific Chemicals Supplier No. H6207103

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 445303-14-0
Molecular Formula C13H15BF4O2
Molecular Weight (g/mol) 290.065
MDL Number MFCD09878546
InChI Key JGMQFPUMJCFTAR-UHFFFAOYSA-N
Synonym 4-fluoro-3-trifluoromethyl phenylboronic acid pinacol ester, 2-4-fluoro-3-trifluoromethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-fluoro-3-trifluoromethyl phenylboronicacidpinacolester, 4-fluoro-3-trifluoromethyl benzeneboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-4-fluoro-3-trifluoromethyl phenyl-4,4,5,5-tetramethyl, 2-4-fluoro-3-trifluoromethyl-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2-4-fluoro-3-trifluoromethylphenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane
PubChem CID 22240176
IUPAC Name 2-[4-fluoro-3-(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)C(F)(F)F

Specifications

Quantity 1 g
Formula Weight 290.06
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)benzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.