Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-4'-hydroxybiphenyl 98.0+%, TCI America™
SDP

Catalog No. F070425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F070425G 25 g
F07045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F070425G Supplier TCI America Supplier No. F070425G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 324-94-7
Molecular Formula C12H9FO
Molecular Weight (g/mol) 188.201
MDL Number MFCD01830385
InChI Key QSJNKJGPJVOGPK-UHFFFAOYSA-N
Synonym 4'-fluoro-1,1'-biphenyl-4-ol, 4-fluoro-4'-hydroxybiphenyl, 4-hydroxy-4'-fluorobiphenyl, 4-4-fluorophenyl phenol, 4'-fluoro-biphenyl-4-ol, 4'-fluoro 1,1'-biphenyl-4-ol, 4-4'-fluorophenyl phenol, 4'-fluorobiphenyl-1-ol, 1,1'-biphenyl-4-ol, 4'-fluoro, pubchem9057
PubChem CID 345420
IUPAC Name 4-(4-fluorophenyl)phenol
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)F)O

Specifications

Melting Point 172°C
Color White-Yellow
Quantity 25 g
Formula Weight 188.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-4′-hydroxybiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.