Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-alpha-methylstyrene, 95%

Catalog No. AAA1414718
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1414718 50 g
AAA1414709 10 g
AAA1414730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1414718 Supplier Thermo Scientific Chemicals Supplier No. A1414718
Add to Cart

CAS: 350-40-3 | C9H9F | 136.169 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 350-40-3
Molecular Formula C9H9F
Molecular Weight (g/mol) 136.169
MDL Number MFCD00042297
InChI Key VIXHMBLBLJSGIB-UHFFFAOYSA-N
Synonym 4-fluoro-alpha-methylstyrene, 1-fluoro-4-1-methylethenyl benzene, 1-fluoro-4-prop-1-en-2-yl benzene, 2-4-fluorophenyl-1-propene, 2-4-fluorophenyl propene, benzene, 1-fluoro-4-1-methylethenyl, 4-fluoro-.alpha.-methylstyrene, 1-fluoro-4-isopropenyl benzene, 1-fluoro-4-isopropenylbenzene, p-fluoro-.alpha.-methylstyrene
PubChem CID 67690
IUPAC Name 1-fluoro-4-prop-1-en-2-ylbenzene
SMILES CC(=C)C1=CC=C(C=C1)F

Specifications

Density 1.009
Boiling Point 183°C
Flash Point 53°C (127°F)
Refractive Index 1.511
Quantity 50 g
UN Number UN1993
Beilstein 2039696
Formula Weight 136.17
Percent Purity 95%
Chemical Name or Material 4-Fluoro-alpha-methylstyrene
Hazard Category H226
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 206-501-2
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.