Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-DL-mandelic Acid 96.0+%, TCI America™
SDP

Catalog No. F090725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F090725G 25 g
F09075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F090725G Supplier TCI America Supplier No. F090725G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 395-33-5
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.14
MDL Number MFCD00044402,MFCD20638902
InChI Key RWCMOQXHIDWDDJ-UHFFFAOYNA-N
Synonym 4-fluoromandelic acid, 2-4-fluorophenyl-2-hydroxyacetic acid, p-fluoro mandelic acid, 4-fluorophenyl hydroxy acetic acid, p-fluoromandelic acid, 4-fluoro mandelic acid, 4-fluoromandelicacid, 4-fluoro-alpha-hydroxybenzeneacetic acid, 4-fluoro-dl-mandelic acid, 4-fluoro hydroxy acetic acid
PubChem CID 98068
IUPAC Name 2-(4-fluorophenyl)-2-hydroxyacetic acid
SMILES OC(C(O)=O)C1=CC=C(F)C=C1

Specifications

Melting Point 137°C
Color White
Quantity 25 g
Formula Weight 170.14
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-DL-mandelic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.