Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-L-phenylalanine, 98+%

Catalog No. AAL1993406
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1993406 5 g
AAL19934MD 250 mg
AAL1993403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1993406 Supplier Thermo Scientific Chemicals Supplier No. L1993406
Add to Cart

CAS: 1132-68-9 | C9H10FNO2 | 183.182 g/mol

4-Fluoro-L-phenylalanine is a substrate for tyrosine hydroxylase (TH) that has been used to study the regulation of that enzyme.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-L-phenylalanine is a substrate for tyrosine hydroxylase (TH) that has been used to study the regulation of that enzyme.

Solubility
Soluble in water and 0.5M HCl (50 mg/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1132-68-9
Molecular Formula C9H10FNO2
Molecular Weight (g/mol) 183.182
MDL Number MFCD00063064
InChI Key XWHHYOYVRVGJJY-QMMMGPOBSA-N
Synonym 4-fluoro-l-phenylalanine, p-fluoro-l-phenylalanine, s-4-fluorophenylalanine, l-4-fluorophenylalanine, h-phe 4-f-oh, s-2-amino-3-4-fluorophenyl propanoic acid, unii-e2k2vdk6kk, l-phenylalanine, 4-fluoro, s---p-fluorophenylalanine, 2s-2-amino-3-4-fluorophenyl propanoic acid
PubChem CID 716312
ChEBI CHEBI:44909
IUPAC Name (2S)-2-amino-3-(4-fluorophenyl)propanoic acid
SMILES C1=CC(=CC=C1CC(C(=O)O)N)F

Specifications

Melting Point ∽240°C (decomposition)
Beilstein 2416148
Solubility Information Soluble in water and 0.5M HCl (50mg/ml).
Quantity 5 g
Formula Weight 183.18
Optical Rotation −25° (c=1 in Water)
Percent Purity ≥98%
Chemical Name or Material 4-Fluoro-L-phenylalanine
RTECSNumber DW1765000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.