Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoroaniline 98.0+%, TCI America™
SDP

Catalog No. F0033500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0033500ML 500 mL
F003325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0033500ML Supplier TCI America Supplier No. F0033500ML

Chemical Identifiers

CAS 371-40-4
Molecular Formula C6H6FN
Molecular Weight (g/mol) 111.12
MDL Number MFCD00007829
InChI Key KRZCOLNOCZKSDF-UHFFFAOYSA-N
Synonym p-fluoroaniline, benzenamine, 4-fluoro, 1-amino-4-fluorobenzene, 4-fluorobenzenamine, p-fluorophenylamine, 4-fluoronaniline, aniline, p-fluoro, aniline, 4-fluoro, 4-fluoranilin, para-fluoroaniline
PubChem CID 9731
ChEBI CHEBI:28546
IUPAC Name 4-fluoroaniline
SMILES NC1=CC=C(F)C=C1

Specifications

Melting Point -1°C
Color Yellow
Boiling Point 188°C
Quantity 500 mL
UN Number 2941
Formula Weight 111.12
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Fluoroaniline
EINECSNumber (3)-0222
RTECSNumber BY1575000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.