Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Fluorobenzylamine

Catalog No. 1624935G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-249-35G 5 g
16-249-325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-249-35G Supplier Sigma Aldrich Supplier No. 1624935G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • [18f]4-fluorobenzylamine reacts with the α- and α-carboxyl groups of folic acid to yield 18f-labeled folate
  • It is an important building block for the synthesis of 18f-labeled compounds.4-fluorobenzylamine was used in the synthesis of new tris-iron(iii) chelates of 3-hydroxy-4-pyridinone ligands

Specifications

CAS 140-75-0
Density 1.095 g/mL (at 25°C (literature))
Boiling Point 183°C (lit.)
Molecular Formula C7H8FN
Refractive Index n20/D 1.512 (literature)
Linear Formula FC6H4CH2NH2
MDL Number MFCD00008120
Quantity 5 g
Molecular Weight (g/mol) 125.14
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.