Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorobenzylamine, 97+%, Thermo Scientific™

Catalog No. AA2223718
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA2223718 50 g
AA2223704 2 g
AA2223709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA2223718 Supplier Thermo Scientific Chemicals Supplier No. 2223718

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140-75-0
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.146
MDL Number MFCD00008120
InChI Key IIFVWLUQBAIPMJ-UHFFFAOYSA-N
Synonym 4-fluorobenzylamine, 4-fluorophenyl methanamine, benzenemethanamine, 4-fluoro, p-fluorobenzylamine, benzylamine, p-fluoro, p-fluorobenzyl amine, 4-fluoro-benzylamine, 1-4-fluorophenyl methanamine, 4-fluorobenzyl amine, chembl12392
PubChem CID 67326
IUPAC Name (4-fluorophenyl)methanamine
SMILES C1=CC(=CC=C1CN)F

Specifications

Density 1.095g/mL
Boiling Point 183°C
Flash Point 66° (152°F)
Quantity 50 g
UN Number UN2735
Formula Weight 125.15
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material 4-Fluorobenzylamine
Hazard Category Danger
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.