Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorocatechol 98.0+%, TCI America™
SDP

Catalog No. F04831G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04831G 1 g
F04835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F04831G Supplier TCI America Supplier No. F04831G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 367-32-8
Molecular Formula C6H5FO2
Molecular Weight (g/mol) 128.102
MDL Number MFCD01317539
InChI Key NFWGQJUHSAGJBE-UHFFFAOYSA-N
Synonym 4-Fluoro-1,2-dihydroxybenzene, 4-Fluoropyrocatechol
PubChem CID 160458
ChEBI CHEBI:80515
IUPAC Name 4-fluorobenzene-1,2-diol
SMILES C1=CC(=C(C=C1F)O)O

Specifications

Melting Point 91°C
Color White-Yellow
Quantity 1 g
UN Number 1759
Formula Weight 128.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluorocatechol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.