Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorocinnamic acid, 98+%

Catalog No. p-7023300
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1670706 5 g
AAA1670714 25 g
AAA1670722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1670706 Supplier Thermo Scientific Chemicals Supplier No. A1670706
Only null left
Add to Cart

CAS: 459-32-5 | C9H7FO2 | 166.151 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 459-32-5
Molecular Formula C9H7FO2
Molecular Weight (g/mol) 166.151
MDL Number MFCD00004395
InChI Key ISMMYAZSUSYVQG-ZZXKWVIFSA-N
Synonym 4-fluorocinnamic acid, e-3-4-fluorophenyl acrylic acid, 2e-3-4-fluorophenyl prop-2-enoic acid, p-fluorocinnamic acid, 3-4-fluorophenyl acrylic acid, e-p-fluorocinnamic acid, trans-4-fluorocinnamic acid, p-fluoro cinnamic acid, 3-4-fluorophenyl prop-2-enoic acid, 2-propenoic acid, 3-4-fluorophenyl-, 2e
PubChem CID 1530234
IUPAC Name (E)-3-(4-fluorophenyl)prop-2-enoic acid
SMILES C1=CC(=CC=C1C=CC(=O)O)F

Specifications

Melting Point 208°C to 210°C
Odor Odorless
Quantity 5 g
Beilstein 2613441
Formula Weight 166.15
Percent Purity ≥98%
Chemical Name or Material 4-Fluorocinnamic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-288-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.