Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorophenethyl bromide, 97%

Catalog No. AC398050050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC398050050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC398050050 Supplier Thermo Scientific Chemicals Supplier No. 398050050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 332-42-3 | C8H8BrF | 203.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 332-42-3
Molecular Formula C8H8BrF
Molecular Weight (g/mol) 203.05
MDL Number MFCD03844794
InChI Key FLRUNCJXOVYWDH-UHFFFAOYSA-N
Synonym 1-2-bromoethyl-4-fluorobenzene, 1-fluoro-4-2-bromoethyl benzene, 4-fluorophenethyl bromide, 2-4-fluorophenyl ethyl bromide, 1-bromo-2-4-fluorophenyl ethane, 2-4-fluorophenyl ethylbromide, 4-fluorophenylethyl bromide, 1-2-bromo-ethyl-4-fluoro-benzene, benzene, 1-2-bromoethyl-4-fluoro, 1-2-bromoethyl-4-fluoro-benzene
PubChem CID 573153
IUPAC Name 1-(2-bromoethyl)-4-fluorobenzene
SMILES FC1=CC=C(CCBr)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow or White to Yellow
Density 1.4498g/mL
Boiling Point 100°C to 104°C (15.0 mmHg)
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5310 to 1.5350 (20°C, 589nm)
Linear Formula FC6H4(CH2)2Br
Quantity 5 g
Specific Gravity 1.4498
Formula Weight 203.05
Percent Purity 97%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 4-Fluorophenethyl bromide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.