Learn More
Sigma Organic Chemistry 4-Fluorophenyl pyridin- 1G
Supplier: Sigma Organic Chemistry OTV0006081G

4-Fluorophenyl pyridin- , 1G
About this item:
MDL #: MFCD15142050
Chemical Formula: C12H12ClFN2
Molecular Weight: 238.69
UNSPSC Code: 12352200
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.