Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Fluoropropiophenone 97.0+%, TCI America™
SDP

Catalog No. F052225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F052225G 25 g
F05225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F052225G Supplier TCI America Supplier No. F052225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 456-03-1
Molecular Formula C9H9FO
Molecular Weight (g/mol) 152.168
InChI Key QIJNVLLXIIPXQT-UHFFFAOYSA-N
Synonym 4'-fluoropropiophenone, 1-4-fluorophenyl propan-1-one, p-fluoropropiophenone, 4-fluoropropiophenone, ethyl 4-fluorophenyl ketone, 1-propanone, 1-4-fluorophenyl, propiophenone, 4'-fluoro, para-fluoropropiophenone, 1-4-fluorophenyl-1-propanone, 4;-fluoropropiophenone
PubChem CID 68004
IUPAC Name 1-(4-fluorophenyl)propan-1-one
SMILES CCC(=O)C1=CC=C(C=C1)F

Specifications

Color Yellow
Boiling Point 210°C
Quantity 25 g
Formula Weight 152.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4′-Fluoropropiophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.