Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Formyl-trans-stilbene 98.0+%, TCI America™
SDP

Catalog No. F041225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F041225G 25 g
F04125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F041225G Supplier TCI America Supplier No. F041225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 40200-69-9
Molecular Formula C15H12O
Molecular Weight (g/mol) 208.26
MDL Number MFCD00011580
InChI Key CLXSBHRRZNBTRT-VOTSOKGWSA-N
Synonym trans-4-Stilbenecarboxaldehyde
PubChem CID 5375876
IUPAC Name 4-[(E)-2-phenylethenyl]benzaldehyde
SMILES C1=CC=C(C=C1)C=CC2=CC=C(C=C2)C=O

Specifications

Melting Point 114°C
Color Yellow
Quantity 25 g
Formula Weight 208.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Formyl-trans-stilbene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.