Learn More
Labconco™ 4 ft. Logic™ Class II, Type B2 Biosafety Cabinet, US Model

Total exhaust cabinets that provide personnel, product and environmental protection from hazardous particulates requiring BSL 1-4 containment. These models require a dedicated exhaust system and remote blower. All models are NSF/ANSI 49 listed and maximize safety, performance, and comfort.
$22525.00 - $24430.00
Specifications
Description | 4 ft. Logic Class II, Type B2 Biosafety Cabinet |
---|---|
Certifications/Compliance | NSF/ANSI 49, ETL, UL, CAN/CSA, ADA, ISO 5 Conditions per ISO 14644-1 and 2 |
Chamber Material | Stainless Steel (304) |
Width (Metric) Interior | 123.2 cm |
Product Type | Class II, Type B2 Bio Safety Cabinet |
Catalog Number | Mfr. No. | Plug Type | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Plug Type | Includes | Price | Quantity & Availability | ||||
01-812-561
![]() |
Labconco™
303480002 |
North America (115 V, 15 A) | Cabinet |
Each for $22,525.00
|
|
||||
01-812-559
![]() |
Labconco™
303480102 |
North America (115 V, 15 A) | Cabinet, Base Stand |
Each for $23,480.00
|
|
||||
01-812-557
![]() |
Labconco™
303481002 |
North America (115 V, 15 A) | Cabinet, Accessory Package |
Each for $23,470.00
|
|
||||
01-812-555
![]() |
Labconco™
303481102 |
North America (115 V, 15 A) | Cabinet, Accessory Package, Base Stand |
Each for $24,430.00
|
|
||||
01-812-560
![]() |
Labconco™
303480012 |
North America (230 V) | Cabinet |
Each for $22,525.00
|
|
||||
01-812-558
![]() |
Labconco™
303480112 |
North America (230 V) | Cabinet, Base Stand |
Each for $23,480.00
|
|
||||
01-812-556
![]() |
Labconco™
303481012 |
North America (230 V) | Cabinet, Accessory Package |
Each for $23,470.00
|
|
||||
01-812-554
![]() |
Labconco™
303481112 |
North America (230 V) | Cabinet, Accessory Package, Base Stand |
Each for $24,430.00
|
|
||||
Description
Performance Features:
- Safe Class II airflows (100% exhaust, remote blower required) that provide personnel, product, and environmental protection.
- Cabinet interior provides ISO Class 5 (Grade A) or better working conditions.
- Constant Airflow Profile™ (CAP) and BalanceFlow™ technology maintain safe downflow – even as HEPA filters load.
- Optimized airflow minimizes energy use and noise output to industry-leading levels.
- Logic OS with 5 in. line-of-sight touchscreen display.
- Access cabinet information such as filter life, alerts and alarms, data logs.
- Remote connectivity, particle counter integration, and certification records elevate user control of the cabinet.
- Smart-Start™ and Night-Smart™ modes simplify startup and shut-down procedures to help ensure consistency and cleanliness before, during, and after work.
- Inclination Technology™ improves ergonomic working conditions by maximizing comfort and accessibility during long periods of work.
- Remote Communication Board for direct communication with remote blower and/or building management system featuring dry contact relays, USB data output and RS-232 RJ45 data output.
- Mag-Zone™ area and mating accessories keep common items such as pipettes and biohazard waste organized and within reach.
- Full, six-year warranty on all Class II biosafety cabinets.
Optional Configurations:
- Accessory Package: UV lamp (254 nm), right-side service fixture with ball-type valve and right and left-side Vacu-Pass™ Cord and Cable Portals
- Telescoping base stand with shelf and leveling feet
Specifications
4 ft. Logic Class II, Type B2 Biosafety Cabinet | |
Stainless Steel (304) | |
Class II, Type B2 Bio Safety Cabinet | |
61 dBA | |
105 W | |
72.7 in | |
54.4 in. | |
5 in. touchscreen | |
HEPA | |
184.7 cm | |
818 CFM | |
48.5 in. | |
22.5 in. | |
65.3 cm | |
69 cm | |
81.3 cm |
NSF/ANSI 49, ETL, UL, CAN/CSA, ADA, ISO 5 Conditions per ISO 14644-1 and 2 | |
123.2 cm | |
Logic B2 | |
2 Duplex | |
2 Double, Left and Right Side | |
20 cm | |
1390 m³/hr. | |
99.99% | |
138.2 cm | |
8 in. | |
6 Years | |
25.7 in. | |
27 in. | |
57.2 cm | |
32 in. | |
105 W |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.