Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Labconco™ 4 ft. Purifier Axiom Class II Type C1 Biosafety Cabinet with 10 in. Sash Opening

4 ft. Purifier Axiom Class II Type C1 Biosafety Cabinet with 10 in. Sash Opening is an unique, flexible BSC that can be operated as Type A or B and is the only Type C1 in the world.
$20480.00 - $22340.00
Specifications
Description | Class II Type C1 Biosafety Cabinet |
---|---|
Certifications/Compliance | NSF/ANSI 49, UL 61010, CAN/CSA C22.2, CE Marked, ISO 5 Conditions per ISO 14644-1 and 2, ADA |
Chamber Material | Stainless Steel |
Product Type | Class II Biosafety Cabinet |
Model | Axiom |
Catalog Number | Mfr. No. | Plug Type | Electrical Requirements | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Plug Type | Electrical Requirements | Includes | Price | Quantity & Availability | |||
30-441-0021
![]() |
Labconco™
304410021 |
Japan | 100 V, 50/60 Hz, 12 A | Biosafety Cabinet Only |
Each for $20,480.00
|
|
|||
30-441-0031
![]() |
Labconco™
304410031 |
Schuko | 230 V, 50/60 Hz, 6 A | Biosafety Cabinet Only |
Each for $20,480.00
|
|
|||
30-441-0041
![]() |
Labconco™
304410041 |
China, Australia | 230 V, 50/60 Hz, 6 A | Biosafety Cabinet Only |
Each for $20,480.00
|
|
|||
30-441-0051
![]() |
Labconco™
304410051 |
UK | 230 V, 50/60 Hz, 6 A | Biosafety Cabinet Only |
Each for $20,480.00
|
|
|||
30-441-0071
![]() |
Labconco™
304410071 |
India | 230 V, 50/60 Hz, 6 A | Biosafety Cabinet Only |
Each for $20,480.00
|
|
|||
30-441-0121
![]() |
Labconco™
304410121 |
Japan | 100 V, 50/60 Hz, 12 A | Base Stand |
Each for $21,420.00
|
|
|||
30-441-0131
![]() |
Labconco™
304410131 |
Schuko | 230 V, 50/60 Hz, 6 A | Base Stand |
Each for $21,420.00
|
|
|||
30-441-0141
![]() |
Labconco™
304410141 |
China, Australia | 230 V, 50/60 Hz, 6 A | Base Stand |
Each for $21,420.00
|
|
|||
30-441-0151
![]() |
Labconco™
304410151 |
UK | 230 V, 50/60 Hz, 6 A | Base Stand |
Each for $21,420.00
|
|
|||
30-441-0171
![]() |
Labconco™
304410171 |
India | 230 V, 50/60 Hz, 6 A | Base Stand |
Each for $21,420.00
|
|
|||
30-441-1021
![]() |
Labconco™
304411021 |
Japan | 100 V, 50/60 Hz, 12 A | Accessory Pk. |
Each for $21,400.00
|
|
|||
30-441-1031
![]() |
Labconco™
304411031 |
Schuko | 230 V, 50/60 Hz, 6 A | Accessory Pk. |
Each for $21,400.00
|
|
|||
30-441-1041
![]() |
Labconco™
304411041 |
China, Australia | 230 V, 50/60 Hz, 6 A | Accessory Pk. |
Each for $21,400.00
|
|
|||
30-441-1051
![]() |
Labconco™
304411051 |
UK | 230 V, 50/60 Hz, 6 A | Accessory Pk. |
Each for $21,400.00
|
|
|||
30-441-1071
![]() |
Labconco™
304411071 |
India | 230 V, 50/60 Hz, 6 A | Accessory Pk. |
Each for $21,400.00
|
|
|||
30-441-1121
![]() |
Labconco™
304411121 |
Japan | 100 V, 50/60 Hz, 12 A | Base Stand, Accessory Pk. |
Each for $22,340.00
|
|
|||
30-441-1131
![]() |
Labconco™
304411131 |
Schuko | 230 V, 50/60 Hz, 6 A | Base Stand, Accessory Pk. |
Each for $22,340.00
|
|
|||
30-441-1141
![]() |
Labconco™
304411141 |
China, Australia | 230 V, 50/60 Hz, 6 A | Base Stand, Accessory Pk. |
Each for $22,340.00
|
|
|||
30-441-1151
![]() |
Labconco™
304411151 |
UK | 230 V, 50/60 Hz, 6 A | Base Stand, Accessory Pk. |
Each for $22,340.00
|
|
|||
30-441-1171
![]() |
Labconco™
304411171 |
India | 230 V, 50/60 Hz, 6 A | Base Stand, Accessory Pk. |
Each for $22,340.00
|
|
|||
Description
- Requires supporting base if included stand option is not selected
- Intrinsically-safe negative pressure design includes:
- Dual Electronically Commutated Motors (ECM)
- Constant Airflow Profile™ (CAP) Technology airflow monitoring system
- Active Protection Protocol operates the cabinet's exhaust blowers for up to five minutes preventing the flow of air into the laboratory in the event of exhaust system failure
- BSC required Air-Wave™ Entry System at access opening to enhance containment
- Contain-Air™ Negative Pressure Channel at top of sash to enhance containment
- Two programmable working sash heights (8 in. and 10 in.)
- 10° angled, fully-closing, 1/4 in. tempered safety glass sash with counterbalanced, anti-racking, easy-to-lift mechanism
- Curved waterfall design stainless steel inlet grille with Reserve-Air™ Secondary Airflow Slots
- 22.6 in. (58 cm) maximum sash opening height and 27.0 in. (69 cm) viewing height Chem-Zone™ work surface with dedicated direct exhaust for use with hazardous vapors or radionuclides
- Leak-tight interior with removable, type 304 stainless steel 3-piece work surface (dished Chem-Zone in center and two flat sides) with lift out knobs and clearly delineated working area
- Nominal inflow velocity of 105 feet per minute (fpm) (0.53 m/sec)
- Nominal downflow velocity of 65 fpm (0.33 m/sec) for 4 ft. models and 55 fpm (0.28 m/sec) for 6 ft. models
- Touchpad on right-hand side post controls blower, light(s), timer, electrical receptacles, audible alarm mute and menu selection
- Omni-Flex™ two mode design (Type A and B mode)
- Low static pressure and volumetric rate exhaust requirements when in Type B mode
- Night-Smart™ System for idling the blower when the sash is fully closed (while in Type A mode only)
- MyLogic™ Operating System and interior-mounted, line-of-sight, full color, easy-to-understand LCD information center with:
- Smart-Start™ System for start up and shut down operations
- Filter Life Remaining bar graph for each HEPA filter
- Status line for alarm conditions and alerts to warn when filter life diminishes to 20% and 0%
- Bright, 90-150 ft. candle, glare-free LED lighting located outside the contaminated work area
- Interval/elapsed timer for experiment monitoring, LED light or UV light control (on models with UV light)
- Electronic security lock (optional activation) that requires access code to operate the cabinet
Specifications
Class II Type C1 Biosafety Cabinet | |
Stainless Steel | |
Axiom | |
2 | |
2 | |
54.3 in. | |
Full-color LCD | |
HEPA | |
Powder Coated Steel | |
10 in. | |
64.2 in. | |
32.7 in. | |
636 lb. |
NSF/ANSI 49, UL 61010, CAN/CSA C22.2, CE Marked, ISO 5 Conditions per ISO 14644-1 and 2, ADA | |
Class II Biosafety Cabinet | |
67 dBA | |
360 W | |
25 cm | |
680 m3/hr. | |
0.9999 | |
137.9 cm | |
LED | |
5 Years | |
163.1 cm | |
83.1 cm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title