Learn More
Container & Packaging Supply 4 gallon white HDPE plastic square pail lid with tear-tab (no gasket) of 55 mil thickness
SDP
Supplier:  Container & Packaging Supply PL104WT
Encompass
The PL104WT is a 4 gallon white HDPE plastic square pail lid with tear-tab (no gasket). This is a heavy-duty pail lid and is ideal for industrial, heavy-duty applications. When fastened to a pail, these filled pails can be stacked on top of each other to optimize storage space. This lid has been equipped with a tear-tab tamper-evident mechanism. Once the lid has been initially fastened to the pail, it cannot be removed without removing the tear-tab. High density polyethylene (HDPE) is marked with the resin identification code of 2. HDPE has a high tensile strength, good resistance to chemicals, good impact resistance, highly rigid, and highly scratch resistant.
Catalog No. NC1624268
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.