Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Glycidyloxycarbazole 98.0+%, TCI America™
SDP

Catalog No. G029625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G029625G 25 g
G02965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G029625G Supplier TCI America Supplier No. G029625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 51997-51-4
Molecular Formula C15H13NO2
Molecular Weight (g/mol) 239.274
MDL Number MFCD03411884
InChI Key SVWKIGRDISDRLO-UHFFFAOYSA-N
Synonym 4-(2,3-Epoxypropoxy)carbazole
PubChem CID 5020710
IUPAC Name 4-(oxiran-2-ylmethoxy)-9H-carbazole
SMILES C1C(O1)COC2=CC=CC3=C2C4=CC=CC=C4N3

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 239.27
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Glycidyloxycarbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.