Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Heptanol 97.0+%, TCI America™
SDP

Catalog No. H0036100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0036100ML 100 mL
H003625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0036100ML Supplier TCI America Supplier No. H0036100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 589-55-9
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.204
MDL Number MFCD00021934
InChI Key YVBCULSIZWMTFY-UHFFFAOYSA-N
Synonym 4-heptanol, dipropylcarbinol, unii-yg7b8091bp, di-n-propylcarbinol, heptanol-4, 4-heptyl alcohol, n-heptan-4-ol, acmc-209m8k, yvbculsizwmtfy-uhfffaoysa, ch3 ch2 2choh ch2 2ch3
PubChem CID 11513
IUPAC Name heptan-4-ol
SMILES CCCC(CCC)O

Specifications

Melting Point -42°C
Color Colorless
Boiling Point 155°C
Quantity 100 mL
UN Number 1987
Formula Weight 116.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Heptanol
EINECSNumber (2)-0217
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.