Learn More
4-Heptanone, 98%
Description
4-Heptanone is used as a solvent for nitrocellulose as well as in organic synthesis. It is involved in the preparation of flavorings. It is used as an intermediate in the synthesis of active pharmaceutical ingredients. Further, it is used in paints and coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Heptanone is used as a solvent for nitrocellulose as well as in organic synthesis. It is involved in the preparation of flavorings. It is used as an intermediate in the synthesis of active pharmaceutical ingredients. Further, it is used in paints and coatings.
Solubility
Miscible with alcohol, ligroin and ether. Immiscible with water.
Notes
Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
Specifications
Specifications
| Melting Point | -33°C |
| Density | 0.818 |
| Boiling Point | 143°C to 144°C |
| Flash Point | 48°C (118°F) |
| Refractive Index | 1.407 |
| Quantity | 250 mL |
| UN Number | UN2710 |
| Beilstein | 1699049 |
| Merck Index | 14,3345 |
| Solubility Information | Miscible with alcohol,ligroin and ether. Immiscible with water. |
| Show More |
Safety and Handling
| Hazard Category | H226-H332 |
| Hazard Statement | GHS H Statement H226-H332 Flammable liquid and vapour. Harmful if inhaled. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: DIPROPYL KETONE |
| EINECSNumber | 204-608-9 |
| RTECSNumber | MJ5600000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.